Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 143 publications

$268.00 - $562.50

Specifications

Antigen ABCG1
Concentration 1 mg/mL
Dilution Western Blot 1:500, Flow Cytometry, Immunohistochemistry 1:100-1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23185679), Immunoprecipitation reported in scientific literature (PMID 30235209), Immunohistochemistry-Paraffin 1:100-1:400, Immunoblotting reported in scientific literature (PMID 28264879), In vitro assay, In vivo assay reported in scientific literature, Flow (Intracellular)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In vitro Assay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB400132SS
SDP
View Documents
Novus Biologicals
NB400132SS
0.025 mL
Each for $268.00
Only null left
Add to Cart
 
NB400132
SDP
View Documents
Novus Biologicals
NB400132
0.1 mL
Each for $562.50
Only null left
Add to Cart
 
Description

Description

ABCG1 Polyclonal specifically detects ABCG1 in Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, In vitro assay, Flow (Intracellular).
Specifications

Specifications

ABCG1
Western Blot 1:500, Flow Cytometry, Immunohistochemistry 1:100-1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 23185679), Immunoprecipitation reported in scientific literature (PMID 30235209), Immunohistochemistry-Paraffin 1:100-1:400, Immunoblotting reported in scientific literature (PMID 28264879), In vitro assay, In vivo assay reported in scientific literature, Flow (Intracellular)
Polyclonal
Rabbit
ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
PBS with 0.02% Sodium Azide
ABC transporter 8, ABC8ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), 10ATP-binding cassette transporter member 1 of subfamily G, WHITE1, WHT1
ABCG1
IgG
Affinity Purified
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, In vitro Assay
Unconjugated
RUO
Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit
P45844
9619
A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400). [UniProt# P45844]
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.