Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCG1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400132C
Description
ABCG1 Polyclonal specifically detects ABCG1 in Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular).Specifications
ABCG1 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Flow (Intracellular) | |
P45844 | |
ABCG1 | |
A synthetic peptide made to an internal region of human ABCG1 (between residues 300-400). [UniProt# P45844] | |
0.1 mL | |
Cancer, Lipid and Metabolism, Signal Transduction | |
9619 | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ABC transporter 8, ABC8ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), 10ATP-binding cassette transporter member 1 of subfamily G, WHITE1, WHT1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chinese Hamster, Hamster, Monkey, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction