Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCG1 Rabbit anti-Human, Mouse, Rat, Clone: 4Y7B3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | ABCG1 |
---|---|
Clone | 4Y7B3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ABCG1 Monoclonal antibody specifically detects ABCG1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ABCG1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
ABC Transporters, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
9619 | |
IgG | |
Affinity purified |
4Y7B3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
ABC transporter 8, ABC8, ABCG1, ATP-binding cassette sub-family G member 1, ATP-binding cassette transporter 8, ATP-binding cassette transporter member 1 of subfamily G, ATP-binding cassette, sub-family G (WHITE), member 1, homolog of Drosophila white, MGC34313, White protein homolog, white protein homolog (ATP-binding cassette transporter 8), WHITE1, WHT1 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human ABCG1 (P45844). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title