Learn More
ABCG2/CD338 Antibody (MM0047-2J39), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB11093511FR
Catalog No. NB11093511R
Description
Description
ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
ABCG2/CD338
Monoclonal
DyLight 680
ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Mouse
Protein G purified
RUO
Primary
This was selected for its ability to detect human ABCG2 by flow cytometry and immunocytochemistry.
Store at 4C in the dark.
IgG2
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0047-2J39
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
ABCG2
Human recombinant ABCG2/CD338
0.1 mL
ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Stem Cells
9429
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only