Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCG4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA534855
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534855 Supplier Thermo Scientific Supplier No. PA534855

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7. Predicted reactivity: Mouse (100%), Rat (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May be involved in macrophage lipid homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCG4
Applications Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Abcg4
Gene Accession No. Q9H172
Gene Alias 6430517O04Rik; ABCG4; ATP binding cassette subfamily G member 4; ATP-binding cassette sub-family G member 4; ATP-binding cassette, subfamily G (WHITE), member 4; ATP-binding cassette, sub-family G (WHITE), member 4; putative ABC transporter; WHITE2
Gene Symbols Abcg4
Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 1 and 201 of ABCG4 (Uniprot ID#Q9H172)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.