Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCG5 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306128AF700
Description
ABCG5 Polyclonal specifically detects ABCG5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ABCG5 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ATP-binding cassette sub-family G member 5, ATP-binding cassette, sub-family G (WHITE), member 5, ATP-binding cassette, subfamily G, member 5, sterolin 1, sterolin-1, STSL | |
Produced in rabbits immunized with E. coli-derived Human ABCG5 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
64240 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction