Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABCG5 Monoclonal Antibody (1B5E10)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531603
Description
MA5-31603 has been tested in indirect ELISA.
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the White subfamily. The protein encoded by this gene functions as a half-transporter to limit intestinal absorption and promote biliary excretion of sterols. It is expressed in a tissue-specific manner in the liver, colon, and intestine. This gene is tandemly arrayed on chromosome 2, in a head-to-head orientation with family member ABCG8. Mutations in this gene may contribute to sterol accumulation and atherosclerosis, and have been observed in patients with sitosterolemia.
Specifications
ABCG5 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9H222 | |
ABCG5 | |
Purified recombinant fragment of human ABCG5 (AA: 306-367) expressed in E. coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1B5E10 | |
Unconjugated | |
ABCG5 | |
Abcg5; ATP binding cassette subfamily G member 5; ATP-binding cassette sub-family G (WHITE) member 5 (sterolin 1); ATP-binding cassette sub-family G member 5; ATP-binding cassette, subfamily G (WHITE), member 5; ATP-binding cassette, sub-family G (WHITE), member 5; ATP-binding cassette, sub-family G (WHITE), member 5 (sterolin 1); ATP-binding cassette, subfamily G, member 5; AW112016; cmp; sterolin 1; sterolin-1; STSL; trac | |
Mouse | |
Protein G | |
RUO | |
64240 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction