Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597127 Supplier Invitrogen™ Supplier No. PA597127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLPMVPFKTK DSPGVFSKLG VLLRRVTRNL VRNKLAVITR LLQNLIMGLF LLFFVLRVRS NVLKGAIQDR VGLLYQFVGA TPYTGMLNAV NLFPVLRAVS D; Positive Samples: HL-60; Cellular Location: Membrane, Multi-pass membrane protein.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the White subfamily. The protein encoded by this gene functions as a half-transporter to limit intestinal absorption and promote biliary excretion of sterols. It is expressed in a tissue-specific manner in the liver, colon, and intestine. This gene is tandemly arrayed on chromosome 2, in a head-to-head orientation with family member ABCG8. Mutations in this gene may contribute to sterol accumulation and atherosclerosis, and have been observed in patients with sitosterolemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCG5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ABCG5
Gene Accession No. Q9H222
Gene Alias Abcg5; ATP binding cassette subfamily G member 5; ATP-binding cassette sub-family G (WHITE) member 5 (sterolin 1); ATP-binding cassette sub-family G member 5; ATP-binding cassette, subfamily G (WHITE), member 5; ATP-binding cassette, sub-family G (WHITE), member 5; ATP-binding cassette, sub-family G (WHITE), member 5 (sterolin 1); ATP-binding cassette, subfamily G, member 5; AW112016; cmp; sterolin 1; sterolin-1; STSL; trac
Gene Symbols ABCG5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human ABCG5 (NP_0718811).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64240
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.