Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCG8 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400110AF350
Description
ABCG8 Polyclonal specifically detects ABCG8 in Human, Mouse (Negative) samples. It is validated for Western Blot.Specifications
ABCG8 | |
Polyclonal | |
Western Blot | |
ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
Rabbit | |
Affinity Purified | |
ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
64241.0 | |
Store at 4°C in the dark. |
Western Blot | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ABCG8 | |
A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProtNo. Q9H221] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction