Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABCG8 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB400110F
Description
ABCG8 Polyclonal specifically detects ABCG8 in Human, Mouse (Negative) samples. It is validated for Immunohistochemistry (Negative).Specifications
ABCG8 | |
Polyclonal | |
Immunohistochemistry (Negative) | |
Q9H221 | |
ABCG8 | |
A synthetic peptide from the N-terminal region of human ABCG8 protein. [UniProt# Q9H221] | |
0.1 mL | |
ABC Transporters, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
64241 | |
Human, Mouse (Negative) | |
IgG |
Immunohistochemistry | |
FITC | |
PBS with 0.05% Sodium Azide | |
ATP-binding cassette sub-family G member 8, ATP-binding cassette, sub-family G (WHITE), member 8, ATP-binding cassette, sub-family G (WHITE), member 8 (sterolin 2), GBD4ATP-binding cassette, subfamily G, member 8, MGC142217, sterolin 2, sterolin-2, STSL | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human. Does not appear to react with mouse. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction