Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABCG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595973 Supplier Invitrogen™ Supplier No. PA595973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGKAAEERG LPKGATPQDT SGLQDRLFSS ESDNSLYFTY SGQPNTLEVR DLNYQVDLAS QVPWFEQLAQ FKMPWTSPSC QNSCELGIQN LSFKVRSGQM LAIIGSSGCG RASLLDVITG RGHGGKIKSG QIWINGQPSS PQLVRKCVAH VRQHNQLLPN LTVRETLAFI AQMRLPRTFS QAQRDKRVED VIAELRLRQC ADTRVGNMYV RGLSGGERRR VSIGVQLLWN PGILILDEPT SGLDSFTAHN LVKTLSRLAK GNRLVLISLH QPRSDIFRLF DLVLLMTSGT PIYLGAAQHM VQYFTAIGYP CPRYSNPADF; Positive Samples: A-549; Cellular Location: Membrane, Multi-pass membrane protein.

ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. ABCG8, expressed at high levels in the liver and intestine, normally cooperates with ABCG5 to limit intestinal absorption and promote biliary excretion of sterols. The mutated form of this transporter can lead to sterol accumulation and atherosclerosis or sitosterolemia, a rare autosomal recessive disorder, characterized by hyperabsorption of sterols and the inability to excrete sterols into bile.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABCG8
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Abcg8
Gene Accession No. Q9H221
Gene Alias 1300003C16Rik; ABCG8; AI114946; ATP binding cassette subfamily G member 8; ATP-binding cassette sub-family G member 8; ATP-binding cassette, subfamily G (WHITE), member 8; ATP-binding cassette, sub-family G (WHITE), member 8; ATP-binding cassette, subfamily G, member 8; GBD4; MGC142217; sterolin 2; Sterolin2; sterolin-2; STSL
Gene Symbols Abcg8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human ABCG8 (NP_0718821).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64241
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.