Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABCG8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104397
Description
Antibody detects endogenous levels of total ABCG8.
ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. ABCG8, expressed at high levels in the liver and intestine, normally cooperates with ABCG5 to limit intestinal absorption and promote biliary excretion of sterols. The mutated form of this transporter can lead to sterol accumulation and atherosclerosis or sitosterolemia, a rare autosomal recessive disorder, characterized by hyperabsorption of sterols and the inability to excrete sterols into bile.
Specifications
ABCG8 | |
Polyclonal | |
Unconjugated | |
Abcg8 | |
1300003C16Rik; ABCG8; AI114946; ATP binding cassette subfamily G member 8; ATP-binding cassette sub-family G member 8; ATP-binding cassette, subfamily G (WHITE), member 8; ATP-binding cassette, sub-family G (WHITE), member 8; ATP-binding cassette, subfamily G, member 8; GBD4; MGC142217; sterolin 2; Sterolin2; sterolin-2; STSL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
155192, 64241, 67470 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P58428, Q9DBM0, Q9H221 | |
Abcg8 | |
A synthesized peptide derived from human ABCG8(Accession Q9H221), corresponding to amino acid residues G289-R339. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction