Learn More
Invitrogen™ Abciximab Recombinant Human Monoclonal Antibody (7E3 (Abciximab))

Description
Specificity: Recognises human CD41.
Specifications
Specifications
| Antigen | Abciximab |
| Applications | Flow Cytometry, Inhibition Assays |
| Classification | Recombinant Monoclonal |
| Clone | 7E3 (Abciximab) |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene Alias | c7E3 |
| Host Species | Human |
| Immunogen | CD41 is the integrin alpha chain 2b forming a fibrinogen receptor in heterodimeric complex with beta 3. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.