Learn More
Thermo Scientific™ ABgene™ Grid-Referenced Cluster Tube Racks
GREENER_CHOICE
Catalog No. AB0977
Thermo Scientific™ ABgene™ Grid-Referenced Cluster Tube Racks contain a lockable lid and silicone tube plugs for security during transport.
  • 96 individual 0.65 or 1.2mL tubes
  • Alphanumeric grid-referencing molded into the rack and lid
  • Temperature range -170°C to +120°C
  • Heavy-duty rack in a microplate format and in accordance with SBS footprint
  • Stackable with or without lid
  • Open bottom to allow full robotic access to micro-tubes—each tube is fully supported to withstand pressure applied by robotic systems
  • Sealing options include MULTISIP™ septum plugs, silicone tube plugs, storage plate cap strips
Ordering Alerts:
Supplied in packs of 10 racks, 96 tubes each
TRUSTED_SUSTAINABILITY
Specifications
Description 1.2mL Grid-Referenced Cluster Tube Rack
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.