Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598393 Supplier Invitrogen™ Supplier No. PA598393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABH1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine. Dioxygenase that repairs alkylated single-stranded DNA and RNA containing 3-methylcytosine by oxidative demethylation. Requires molecular oxygen, alpha-ketoglutarate and iron. May have a role in placental trophoblast lineage differentiation. Has DNA lyase activity and introduces double-stranded breaks at abasic sites. Cleaves both single-stranded DNA and double-stranded DNA at abasic sites, with the greatest activity towards double-stranded DNA with two abasic sites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABH1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALKBH1
Gene Accession No. P0CB42, Q13686
Gene Alias 2700073G19Rik; Abh; ABH1; alkB; alkB homolog 1, histone H2A dioxygenase; alkB, alkylation repair homolog 1; alkB, alkylation repair homolog 1 (E. coli); ALKBH; Alkbh1; Alkylated DNA repair protein alkB homolog 1; alkylation repair, alkB homolog; alpha-ketoglutarate-dependent dioxygenase ABH1; DNA 6mA demethylase; DNA demethylase ALKBH1; DNA lyase ABH1; DNA N6-methyl adenine demethylase; DNA N6-methyl adenine demethylase ALKBH1; DNA oxidative demethylase ALKBH1; hABH; mRNA N(3)-methylcytidine demethylase; Nucleic acid dioxygenase ALKBH1; tRNA N1-methyl adenine demethylase
Gene Symbols ALKBH1
Host Species Rabbit
Immunogen Recombinant Human Alkylated DNA repair protein alkB homolog 1 protein (110-389AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211064, 8846
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.