Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABH2 Rabbit anti-Human, Mouse, Rat, Clone: S05-5H7, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | ABH2 |
---|---|
Clone | S05-5H7 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
ABH2 Monoclonal antibody specifically detects ABH2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
ABH2 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Direct Reversal of DNA Damage | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
121642 | |
IgG | |
Affinity-purified |
S05-5H7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
2OG-Fe(II) oxy DC1, ABH2FLJ99103, alkB, alkylation repair homolog 2 (E. coli), Alkylated DNA repair protein alkB homolog 2, alpha-ketoglutarate-dependent dioxygenase alkB homolog 2, EC 1.14.11.-, MGC90512, Oxy DC1 | |
A synthetic peptide of human ABH2 (Uniprot#: Q6NS38) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title