Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299171AF350
Description
ABHD1 Polyclonal specifically detects ABHD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ABHD1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
abhydrolase domain containing 1, EC 3.1.1, EC 3.1.1.-, FLJ36128, LABH1abhydrolase domain-containing protein 1, Lung alpha/beta hydrolase 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ABHD1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
84696 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction