Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ABHD1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ABHD1 Polyclonal specifically detects ABHD1 in Human samples. It is validated for Western Blot.Specifications
| ABHD1 | |
| Polyclonal | |
| Rabbit | |
| NP_115993 | |
| 84696 | |
| Synthetic peptide directed towards the middle region of human ABHD1The immunogen for this antibody is ABHD1. Peptide sequence GLVAALTLSACWDSFETTRSLETPLNSLLFNQPLTAGLCQLVERNRKVIE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| abhydrolase domain containing 1, EC 3.1.1, EC 3.1.1.-, FLJ36128, LABH1abhydrolase domain-containing protein 1, Lung alpha/beta hydrolase 1 | |
| ABHD1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title