Learn More
Invitrogen™ ABHD1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ABHD1 protein.
Specifications
Specifications
| Antigen | ABHD1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ABHD1 |
| Gene Accession No. | Q96SE0 |
| Gene Alias | Abhd1; abhydrolase domain containing 1; abhydrolase domain-containing protein 1; alpha/beta hydrolase domain containing protein 1; alpha/beta hydrolase domain-containing protein 1; alpha/beta hydrolase-1; LABH1; LABH-1; Lung alpha/beta hydrolase 1; mmLABH1; Protein ABHD1; testicular tissue protein Li 5 |
| Gene Symbols | ABHD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.