Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560697
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560697 Supplier Invitrogen™ Supplier No. PA560697
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLEPHCSITT ETFYPTLWCF EGRLQSIFQV LLQSQPLVLY QSDILQTPDG GQLLLDWAKQ PDSSQDPDPT TQPIVLLLPG ITGSSQETYV LHLVNQALRD GYQAVVFNNR GCRGEELRTH R Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 80%.

This gene is a member of the AB hydrolase superfamily and encodes a protein with an alpha/beta hydrolase fold. This domain is common to a number of hydrolytic enzymes of widely differing phylogenetic origins and catalytic functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD1
Gene Accession No. Q96SE0
Gene Alias Abhd1; abhydrolase domain containing 1; abhydrolase domain-containing protein 1; alpha/beta hydrolase domain containing protein 1; alpha/beta hydrolase domain-containing protein 1; alpha/beta hydrolase-1; LABH1; LABH-1; Lung alpha/beta hydrolase 1; mmLABH1; Protein ABHD1; testicular tissue protein Li 5
Gene Symbols ABHD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHD1. Recombinant protein control fragment (Product #RP-90308).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.