Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | ABHD10 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ABHD10 Polyclonal specifically detects ABHD10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ABHD10 | |
Polyclonal | |
Rabbit | |
Proteases & Other Enzymes | |
abhydrolase domain containing 10, EC 3.4, FLJ11342, mitochondrial | |
ABHD10 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
55347 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:STLGKWRKDVLSIIDDLADGPQILVGSSLGGWLMLHAAIARPEKVVALIGVATAADTLVTKFNQLPVELKKEVEMKGVWSM | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title