Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111610
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111610 Supplier Invitrogen™ Supplier No. PA5111610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STLGKWRKDV LSIIDDLADG PQILVGSSLG GWLMLHAAIA RPEKVVALIG VATAADTLVT KFNQLPVELK KEVEMKGVWS M Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 78%.

This gene encodes a mitochondrially-localized enzyme that acts in liver cells as a hydrolase. The encoded protein removes glucuronide from mycophenolic acid acyl-glucuronide. There is a pseudogene for this gene on chromosome 6. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD10
Gene Accession No. Q9NUJ1
Gene Alias Abhd10; abhydrolase domain containing 10; abhydrolase domain containing 10, depalmitoylase; Abhydrolase domain-containing protein 10; abhydrolase domain-containing protein 10, mitochondrial; Alpha/beta hydrolase domain-containing protein 10; alpha/beta hydrolase domain-containing protein 10, mitochondrial; mycophenolic acid acyl-glucuronide esterase, mitochondrial; RGD1308084
Gene Symbols ABHD10
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ABHD10. Recombinant protein control fragment (Product #RP-107475).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55347
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.