Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD11 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299129MFV450
Description
ABHD11 Polyclonal antibody specifically detects ABHD11 in Human samples. It is validated for Western BlotSpecifications
ABHD11 | |
Polyclonal | |
Western Blot | |
abhydrolase domain containing 11, abhydrolase domain-containing protein 11, EC 3.-, PP1226, WBSCR21, Williams Beuren syndrome chromosome region 21, Williams-Beuren syndrome chromosomal region 21 protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ABHD11. | |
0.1 mL | |
Lipid and Metabolism | |
83451 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction