Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555445
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555445 Supplier Invitrogen™ Supplier No. PA555445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALILESPFTN IREEAKSHPF SVIYRYFPGF DWFFLDPITS SGIKFANDEN VKHISCPLLI LHAEDDPVVP FQLGRKLYSI AAPARSFRDF KVQFVPFHSD LGYRHKYIYK SPELPRILRP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

This gene encodes an enzyme that catalyzes the hydrolysis of 2-arachidonoyl glycerol, the main endocannabinoid lipid transmitter that acts on cannabinoid receptors, CB1 and CB2. The endocannabinoid system is involved in a wide range of physiological processes, including neurotransmission, mood, appetite, pain appreciation, addiction behavior, and inflammation. Mutations in this gene are associated with the neurodegenerative disease, PHARC, resulting from an inborn error of endocannabinoid metabolism. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD12
Gene Accession No. Q8N2K0
Gene Alias 1500011G07Rik; 2-arachidonoylglycerol hydrolase; 2-arachidonoylglycerol hydrolase ABHD12; 6330583M11Rik; Abhd12; ABHD12A; abhydrolase domain containing 12; abhydrolase domain-containing protein 12; AI431047; AW547313; BEM46L2; C20orf22; dJ965G21.2; DKFZP434P106; hABHD12; Lysophosphatidylserine lipase ABHD12; Monoacylglycerol lipase ABHD12; Oxidized phosphatidylserine lipase ABHD12; PHARC
Gene Symbols ABHD12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHD12. Recombinant protein control fragment (Product #RP-90030).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26090
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.