Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551760 Supplier Invitrogen™ Supplier No. PA551760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGTGVATNAA KVLEEKGCPV DAIVLEAPFT NMWVASINYP LLKIYRNIPG FLRTLMDALR KDKIVFPNDE NVKFLSSPLL ILHGEDDRTV PLEYGKKLYE IARNAYRNKE RVKMVIFPPG FQHNLLCKSP TLLITVRDFL SKQ Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 44%.

ABHD12B gene ontology annotations related to this gene include hydrolase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD12B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD12B
Gene Accession No. Q7Z5M8
Gene Alias ABHD12B; abhydrolase domain containing 12B; abhydrolase domain-containing protein 12B; Alpha/beta hydrolase domain-containing protein 12B; Bem46l3; c14_5314; C14orf29; Gm804; Protein ABHD12B; serine protease BEM46-like 3; Variant 4
Gene Symbols ABHD12B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHD12B. Recombinant protein control fragment (Product #RP-89048).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145447
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.