Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556944 Supplier Invitrogen™ Supplier No. PA556944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSEGEASEEG LYLDSEAVLD YVMTRPDLDK TKIFLFGRSL GGAVAIHLAS ENSHRISAIM VENTFLSIPH MASTLFSFFP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The α/β hydrolase superfamily comprise diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical α/β hydrolase fold, which consists of an eightstranded parallel α/β structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. The Ab hydrolase domain containing (ABHD) gene subfamily is comprised of 15 mostly uncharacterized members. Most of which utilize a serine nucleophile to form the G-X-S-X-G nucleophile elbow. ABHD1 plays a role in metabolizing smoking xenobiotics. ABHD2 participates in the development of atherosclerosis. ABHD4 is involved in an alternative synthesis pathway of NAE. Mutations in ABHD5 contribute to Chanarin-Dorfman syndrome. ABDH6 may play a role in nervous system metabolism and signaling. ABHD13 is a 337 amino acid single-pass membrane protein that belongs to the ABHD family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD13
Gene Accession No. Q7L211
Gene Alias 1110065L07Rik; Abhd13; abhydrolase domain containing 13; abhydrolase domain-containing protein 13; AI463703; AI788994; Alpha/beta hydrolase domain-containing protein 13; bA153I24.2; BEM46L1; C13orf6; Protein ABHD13; RGD1308317; RP11-153I24.2
Gene Symbols ABHD13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHD13. Recombinant protein control fragment (Product #RP-95842).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84945
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.