Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD14A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546077
Description
Peptide sequence: DLPGFGNSAP SKEASTEAGR AALLERALRD LEVQNAVLVS PSLSGHYALP Sequence homology: Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 86%.
ABHD14A play a possible role in granule neuron development.
Specifications
| ABHD14A | |
| Polyclonal | |
| Unconjugated | |
| ABHD14A | |
| 1110013B16Rik; ABHD14A; abhydrolase domain containing 14A; abhydrolase domain-containing protein 14A; abhydrolase domain-containing protein 14A; protein ABHD14A; alpha/beta hydrolase domain-containing protein 14A; AW558221; DORZ1; down-regulated in Zic1 deficient cerebellar primordium; downregulated in Zic1 deficient cerebellum; down-regulated in Zic-1-mutant protein; protein ABHD14A; RGD1309721; UNQ1913/PRO4373 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25864 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BUJ0 | |
| ABHD14A | |
| Synthetic peptide directed towards the middle region of human ABHD14A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction