Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546078 Supplier Invitrogen™ Supplier No. PA546078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IAPTSTQNYT QEQFWAVKTP TLILYGELDH ILARESLRQL RHLPNHSVVK Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 86%.

ABHD14A play a possible role in granule neuron development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD14A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ABHD14A
Gene Accession No. Q9BUJ0
Gene Alias 1110013B16Rik; ABHD14A; abhydrolase domain containing 14A; abhydrolase domain-containing protein 14A; abhydrolase domain-containing protein 14A; protein ABHD14A; alpha/beta hydrolase domain-containing protein 14A; AW558221; DORZ1; down-regulated in Zic1 deficient cerebellar primordium; downregulated in Zic1 deficient cerebellum; down-regulated in Zic-1-mutant protein; protein ABHD14A; RGD1309721; UNQ1913/PRO4373
Gene Symbols ABHD14A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ABHD14A (aa 194-243).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25864
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.