Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565207
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565207 Supplier Invitrogen™ Supplier No. PA565207
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PEQTSCLWGDP NVTVLAGLTP GNSPIFYREV LPLNQAHRVE VVLLHGKAFN SHTWEQL Highest antigen sequence identity to the following orthologs - mouse 81%, rat 78%.

ABHD14A play a possible role in granule neuron development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD14A
Applications Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD14A
Gene Accession No. Q9BUJ0
Gene Alias 1110013B16Rik; ABHD14A; abhydrolase domain containing 14A; abhydrolase domain-containing protein 14A; abhydrolase domain-containing protein 14A; protein ABHD14A; alpha/beta hydrolase domain-containing protein 14A; AW558221; DORZ1; down-regulated in Zic1 deficient cerebellar primordium; downregulated in Zic1 deficient cerebellum; down-regulated in Zic-1-mutant protein; protein ABHD14A; RGD1309721; UNQ1913/PRO4373
Gene Symbols ABHD14A
Host Species Rabbit
Immunogen Recombinant Human ABHD14A. Recombinant protein control fragment (Product #RP-104684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.