Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD14B Antibody (002), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290259MFV610
Description
ABHD14B Monoclonal antibody specifically detects ABHD14B in Human samples. It is validated for ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
ABHD14B | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunocytochemistry | |
002 | |
ELISA, Immunocytochemistry/ Immunofluorescence | |
abhydrolase domain containing 14B, abhydrolase domain-containing protein 14B, CCG1-interacting factor B, CIBcell cycle gene 1-interacting factor B, EC 3.-, EC 3.1.11.6, EC 3.1.21.4, MGC15429 | |
Rabbit immunized with purified, recombinant Human ABHD14B (Uniprot#: Q96IU4-1, Met1-Gln210) | |
0.1 mL | |
Proteases & Other Enzymes | |
84836 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction