Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD14B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988147
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8147 Supplier Bioss Supplier No. BS7945R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABHEB has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription.

Specifications

Antigen ABHD14B
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ABHD14B
Gene Accession No. Q96IU4
Gene Alias 1810013B01Rik; Abhd14b; abhydrolase domain containing 14B; Abhydrolase domain-containing protein 14B; Alpha/beta hydrolase domain-containing protein 14B; CCG1-interacting factor B; cell cycle gene 1-interacting factor B; CIB; epididymis secretory protein Li 299; HEL-S-299; Protein ABHD14B
Gene Symbols ABHD14B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ABHD14B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84836
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.