Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553174
Description
Immunogen sequence: TALEDTVDTS RLFRSRSLRE FEEALFCHTK SFPISWDAYW DRNDPLRDVD EAAVPVLCIC SADDPVCGPP DHTLTTELFH SNPYFFLLLS RHGGHCGFLR QEPLPAWSHE VILESFRALT EFFRTEERIK GLSR Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
ABHD15 gene ontology annotations related to this gene include cellular lipid metabolic process.
Specifications
| ABHD15 | |
| Polyclonal | |
| Unconjugated | |
| ABHD15 | |
| 1300007F04Rik; ABHD15; abhydrolase domain containing 15; abhydrolase domain-containing protein 15; abhydrolase domain-containing protein 15; protein ABHD15; alpha/beta hydrolase; alpha/beta hydrolase domain-containing protein 15; alpha/beta hydrolase-1; lung alpha/beta hydrolase 1; protein ABHD15; RGD1304598; UNQ6510/PRO21435 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116236 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXT9 | |
| ABHD15 | |
| Recombinant protein corresponding to Human ABHD15. Recombinant protein control fragment (Product #RP-90423). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction