Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD15 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988148
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8148 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8148 Supplier Bioss Supplier No. BS7946R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABHD15 gene ontology annotations related to this gene include cellular lipid metabolic process.

Specifications

Antigen ABHD15
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ABHD15
Gene Accession No. Q6UXT9
Gene Alias 1300007F04Rik; ABHD15; abhydrolase domain containing 15; abhydrolase domain containing 3-like; Abhydrolase domain-containing protein 15; alpha/beta hydrolase; Alpha/beta hydrolase domain-containing protein 15; alpha/beta hydrolase-1; LOC100341891; lung alpha/beta hydrolase 1; Protein ABHD15; RGD1304598; UNQ6510/PRO21435
Gene Symbols ABHD15
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ABHD15.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116236
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.