Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD17A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140126 Supplier Invitrogen™ Supplier No. PA5140126
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGTLRASSGA PGRWKLHLTE RADFQYSQ Highest antigen sequence indentity to the following orthologs: Rat - 71%, Mouse - 71%.

Hydrolyzes fatty acids from S-acylated cysteine residues in proteins (PubMed:26701913). Has depalmitoylating activity towards NRAS (PubMed:26701913). Has depalmitoylating activity towards DLG4/PSD95 (PubMed:26701913). May have depalmitoylating activity towards MAP6. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD17A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD17A
Gene Accession No. Q96GS6
Gene Alias 1700013O15Rik; Abhd17a; abhydrolase domain containing 17A; abhydrolase domain containing 17A, depalmitoylase; abhydrolase domain-containing protein 17A; abhydrolase domain-containing protein FAM108A; abhydrolase domain-containing protein FAM108A1; alpha/beta hydrolase domain-containing protein 17A; BC005632; C19orf27; D10Bwg1364e; FAM108A; FAM108A1; family with sequence similarity 108, member A; family with sequence similarity 108, member A1; mFLJ00358 protein; MGC138013 protein; protein ABHD17A; RGD1359682; upf0227
Gene Symbols ABHD17A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHD17A. Recombinant protein control fragment (Product #RP-109180).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.