Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD17C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561831
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561831 Supplier Invitrogen™ Supplier No. PA561831
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDAVEVFFSR TARDNRLGCM FV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD17C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD17C
Gene Accession No. Q6PCB6
Gene Alias 2210412D01Rik; ABHD17C; abhydrolase domain containing 17C; abhydrolase domain-containing protein 17C; abhydrolase domain-containing protein FAM108C1; AL023007; alpha/beta hydrolase domain-containing protein 17C; Fam108c; Fam108c1; family with sequence similarity 108, member C; family with sequence similarity 108, member C1; protein ABHD17C
Gene Symbols ABHD17C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHD17C. Recombinant protein control fragment (Product #RP-100875).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58489
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.