Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106585
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106585 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106585 Supplier Invitrogen™ Supplier No. PA5106585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABHD2.

This gene encodes a protein containing an alpha/beta hydrolase fold, which is a catalytic domain found in a very wide range of enzymes. The function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Abhd2
Gene Accession No. P08910, Q9QXM0
Gene Alias 2210009N18Rik; 2-arachidonoylglycerol hydrolase; Abhd2; abhydrolase domain containing 2; abhydrolase domain-containing protein 2; Acetylesterase; alpha/beta hydrolase domain containing protein 2; alpha/beta hydrolase fold protein; HS1-2; LABH2; Labh-2; Lung alpha/beta hydrolase 2; mmLABH2; Monoacylglycerol lipase ABHD2; PHPS1-2; Progesterone-sensitive lipase; Protein PHPS1-2; testicular tissue protein Li 6; Triacylglycerol lipase
Gene Symbols Abhd2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ABHD2(Accession P08910), corresponding to amino acid residues Y256-N306.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11057, 293050, 54608
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.