Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597423
Description
ABHD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene encodes a protein containing an alpha/beta hydrolase fold, which is a catalytic domain found in a very wide range of enzymes. The function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding the same protein.
Specifications
ABHD2 | |
Polyclonal | |
Unconjugated | |
Abhd2 | |
2210009N18Rik; 2-arachidonoylglycerol hydrolase; Abhd2; abhydrolase domain containing 2; abhydrolase domain-containing protein 2; Acetylesterase; alpha/beta hydrolase domain containing protein 2; alpha/beta hydrolase fold protein; HS1-2; LABH2; Labh-2; Lung alpha/beta hydrolase 2; mmLABH2; Monoacylglycerol lipase ABHD2; PHPS1-2; Progesterone-sensitive lipase; Protein PHPS1-2; testicular tissue protein Li 6; Triacylglycerol lipase | |
Rabbit | |
Protein G | |
RUO | |
11057, 293050, 54608 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P08910, Q9QXM0 | |
Abhd2 | |
Recombinant Human Monoacylglycerol lipase ABHD2 protein (31-320AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction