Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502205988
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-5988 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-5988 Supplier Fabgennix Inc Supplier No. ABHD2201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABHD2 Polyclonal antibody specifically detects ABHD2 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ABHD2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Abhd2
Gene Accession No. P08910, Q9QXM0
Gene Alias 2210009N18Rik; 2-arachidonoylglycerol hydrolase; Abhd2; abhydrolase domain containing 2; abhydrolase domain-containing protein 2; Acetylesterase; alpha/beta hydrolase domain containing protein 2; alpha/beta hydrolase fold protein; HS1-2; LABH2; Labh-2; Lung alpha/beta hydrolase 2; mmLABH2; Monoacylglycerol lipase ABHD2; PHPS1-2; Progesterone-sensitive lipase; Protein PHPS1-2; testicular tissue protein Li 6; Triacylglycerol lipase
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 200-250 on mouse a/b hydrolase II protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11057, 293050, 54608
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.