Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD4 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2843780.1ML
Description
ABHD4 Polyclonal specifically detects ABHD4 in Rat samples. It is validated for Western Blot.Specifications
ABHD4 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
ABH4, abhydrolase domain containing 4, abhydrolase domain-containing protein 4, Alpha/beta-hydrolase 4, EC 3.1.1.-, FLJ12816, Lyso-N-acylphosphatidylethanolamine lipase | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat ABHD4 | |
0.1 mL | |
Lipid and Metabolism | |
63874 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction