Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551421 Supplier Invitrogen™ Supplier No. PA551421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADDLEQQSQG WLSSWLPTWR PTSMSQLKNV EARILQCLQN KFLARYVSLP NQNKIWTVTV SPEQNDRTPL VMVHGFGGGV GLWILNMDSL SARRTLHTFD LLGFGRSSRP AFPRDPEGAE DEFVTSIETW RETMGIPSMI LLGHSLGGF Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The α/β hydrolase superfamily is comprised of diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical α/β hydrolase fold, which consists of an eight-stranded parallel α/β structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. ABHD1 plays a role in metabolizing smoking xenobiotics. ABHD2 participates in the development of atherosclerosis. ABHD4 is involved in an alternative synthesis pathway of NAE. ABHD4 is a lysophospholipase selective for N-acyl phosphatidylethanolamine (NAPE) which participates in the biosynthesis of N-acyl ethanolamines. Mutations in ABHD5 contribute to Chanarin-Dorfman syndrome. ABDH6 may play a role in nervous system metabolism and signaling. ABHD14A is possibly involved in granule neuron development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD4
Gene Accession No. Q8TB40
Gene Alias (Lyso)-N-acylphosphatidylethanolamine lipase; (Lyso)-N-acylphosphatidylethanolamine lipase; protein ABHD4; 1110035H23Rik; Abh4; Abhd4; abhydrolase domain containing 4; abhydrolase domain containing 4, N-acyl phospholipase B; abhydrolase domain-containing protein 4; AI429574; Alpha/beta hydrolase domain-containing protein 4; alpha/beta-hydrolase 4; hypothetical protein LOC509896; lyso-N-acylphosphatidylethanolamine lipase; protein ABHD4
Gene Symbols ABHD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHD4. Recombinant protein control fragment (Product #RP-88857).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63874
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.