Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116772
Description
Positive Samples: Rat testis Immunogen sequence: PDFKRKFADF FEDDTISEYI YHCNAQNPSG ETAFKAMMES FGWARRPMLE RIHLIRKDVP ITMIYGSDTW IDTSTGKKVK MQRPDSYVRD MEIKGASHHV YADQPHIFNA VVEEICDSVD.
The α/β hydrolase superfamily is comprised of diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical α/β hydrolase fold, which consists of an eight-stranded parallel α/β structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. ABHD1 plays a role in metabolizing smoking xenobiotics. ABHD2 participates in the development of atherosclerosis. ABHD4 is involved in an alternative synthesis pathway of NAE. ABHD4 is a lysophospholipase selective for N-acyl phosphatidylethanolamine (NAPE) which participates in the biosynthesis of N-acyl ethanolamines. Mutations in ABHD5 contribute to Chanarin-Dorfman syndrome. ABDH6 may play a role in nervous system metabolism and signaling. ABHD14A is possibly involved in granule neuron development.
Specifications
ABHD4 | |
Polyclonal | |
Unconjugated | |
ABHD4 | |
(Lyso)-N-acylphosphatidylethanolamine lipase; (Lyso)-N-acylphosphatidylethanolamine lipase; protein ABHD4; 1110035H23Rik; Abh4; Abhd4; abhydrolase domain containing 4; abhydrolase domain containing 4, N-acyl phospholipase B; abhydrolase domain-containing protein 4; AI429574; Alpha/beta hydrolase domain-containing protein 4; alpha/beta-hydrolase 4; hypothetical protein LOC509896; lyso-N-acylphosphatidylethanolamine lipase; protein ABHD4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
105501, 364380, 63874 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.72 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8TB40, Q8VD66 | |
ABHD4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 223-342 of human ABHD4 (NP_0713432). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction