Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abhd5 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10057850
Description
Abhd5 Polyclonal specifically detects Abhd5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Abhd5 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is not expected to cross-react with ABHD4. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Western Blot 0.2 - 0.6 ug/mL, Immunohistochemistry 3.75 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 3.75 ug/mL, Peptide ELISA Detection limit 1:16000 | |
Q8WTS1 | |
ABHD5 | |
Peptide with sequence C-FPERPDLADQDR corresponding to internal region according to NP_057090.2. | |
0.1 mg | |
Lipid and Metabolism, Lipid Droplets | |
51099 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction