Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abhd5 Antibody (SR1756), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32193150UL
Description
Abhd5 Monoclonal antibody specifically detects Abhd5 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
Abhd5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry | |
SR1756 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:500 | |
abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58 | |
A synthesized peptide derived from human Abhd5 (Uniprot #: Q8WTS1) | |
50 μg | |
Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
51099 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction