Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD5 Recombinant Rabbit Monoclonal Antibody (2K8W9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538176
Description
Positive Samples: HepG2, RD, Rat liver, Rat testis Immunogen sequence: TNRPVYAFDL LGFGRSSRPR FDSDAEEVEN QFVESIEEWR CALGLDKMIL LGHNLGGFLA AAYSLKYPSR VNHLILVEPW GFPERPDLAD QDRPIPVWIR A.
The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.
Specifications
ABHD5 | |
Recombinant Monoclonal | |
1.15 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6QA69, Q8WTS1 | |
ABHD5 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human ABHD5 (Q8WTS1). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
2K8W9 | |
Unconjugated | |
ABHD5 | |
1300003D03Rik; 1-acylglycerol-3-phosphate O-acyltransferase ABHD5; 2010002J10Rik; ABHD5; abhydrolase domain containing 5; abhydrolase domain-containing protein 5; aCGI58; CDS; CGI 58; CGI58; CGI-58; CGI58 protein; IECN2; IECN5; lipid droplet-binding protein CGI-58; MGC8731; NCIE2; Protein CGI-58; truncated abhydrolase domain-containing protein 5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
316122, 51099 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction