Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABHD7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557140
Description
Immunogen sequence: FPHPNVFTEY ILRHPAQLLK SSYYYFFQIP WFPEFMFSIN DFKVLKHLFT SHSTGIGRKG CQLTTEDLEA YIYVF Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
ABHD7 gene ontology (GO) annotation includes hydrolase activity and membrane.
Specifications
ABHD7 | |
Polyclonal | |
Unconjugated | |
EPHX4 | |
ABHD7; abhydrolase domain containing 7; abhydrolase domain-containing protein 7; EH4; EPHX4; Ephxrp; epoxide hydrolase 4; Epoxide hydrolase-related protein; Gm1382 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
253152 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IUS5 | |
EPHX4 | |
Recombinant protein corresponding to Human ABHD7. Recombinant protein control fragment (Product #RP-95704). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction