Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106586 Supplier Invitrogen™ Supplier No. PA5106586
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABHD7.

ABHD7 gene ontology (GO) annotation includes hydrolase activity and membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHX4
Gene Accession No. Q6IE26, Q8IUS5
Gene Alias ABHD7; abhydrolase domain containing 7; abhydrolase domain-containing protein 7; EH4; EPHX4; Ephxrp; epoxide hydrolase 4; Epoxide hydrolase-related protein; Gm1382
Gene Symbols EPHX4
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHX4(Accession Q8IUS5), corresponding to amino acid residues W106-D156.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253152, 289440, 384214
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.