Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111464 Supplier Invitrogen™ Supplier No. PA5111464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGYEVVAPDL AGHGASSAPQ VAAAYTFYAL AEDMRAIFKR YAKKRNVLIG HSYGVSFCTF LAHEYPDLVH KVIMINGGGP TALEPSFCSI Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

The alpha/beta hydrolase superfamily comprise diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical alpha/beta hydrolase fold, which consists of an eight-stranded parallel alpha/beta structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. The alpha/beta hydrolase domain containing (ABHD) gene subfamily is comprised of 15 mostly uncharacterized members, most of which utilize a serine nucleophile to form the G-X-S-X-G nucleophile elbow. ABHD8 is a 439 amino acid protein that belongs to the AB hydrolase superfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD8
Gene Accession No. Q96I13
Gene Alias 0910001L24Rik; AB030191; ABHD8; abhydrolase domain containing 8; abhydrolase domain-containing protein 8; Alpha/beta hydrolase domain-containing protein 8; protein ABHD8
Gene Symbols ABHD8
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ABHD8. Recombinant protein control fragment (Product #RP-105097).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.