Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHDB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554962 Supplier Invitrogen™ Supplier No. PA554962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGKTAMLLAL QRPELVERLI AVDISPVEST GVSHFATYVA AMRAINIADE LPRSRARKLA DEQLSSVIQD MAVRQHLLTN LVEVDG Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 65%.

ABHDB encodes a protein containing an alpha/beta hydrolase fold domain. This protein is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHDB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD11
Gene Accession No. Q8NFV4
Gene Alias ABHD11; abhydrolase domain containing 11; abhydrolase domain-containing protein 11; Alpha/beta hydrolase domain-containing protein 11; PP1226; Protein ABHD11; WBSCR21; Williams Beuren syndrome chromosome region 21; Williams-Beuren syndrome chromosomal region 21 protein
Gene Symbols ABHD11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHDB. Recombinant protein control fragment (Product #RP-92657).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.