Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABHEB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557661
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557661 Supplier Invitrogen™ Supplier No. PA557661
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PICTDKINAA NYASVKTPAL IVYGDQDPMG QTSFEHLKQL PNHRVLIMKG AGHPCYLDKP EEWHTGLLDF L Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

ABHEB has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHEB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD14B
Gene Accession No. Q96IU4
Gene Alias 1810013B01Rik; ABHD14B; abhydrolase domain containing 14B; abhydrolase domain-containing protein 14B; Alpha/beta hydrolase domain-containing protein 14B; CCG1-interacting factor B; cell cycle gene 1-interacting factor B; CIB; epididymis secretory protein Li 299; HEL-S-299; Protein ABHD14B
Gene Symbols ABHD14B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABHEB. Recombinant protein control fragment (Product #RP-96316).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84836
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.