Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABI1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059843
Description
ABI1 Polyclonal specifically detects ABI1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
ABI1 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Abelson interactor 1, ABI-1, abl interactor 1, Abl-binding protein 4, AblBP4, abl-interactor 1, Abl-interactor protein 1 long, E3B1, eps8 binding protein, Eps8 SH3 domain-binding protein, Eps8-binding protein, interactor protein AblBP4, nap1 binding protein, Nap1-binding protein, Nap1BP, spectrin SH3 domain binding protein 1, Spectrin SH3 domain-binding protein 1, SSH3BP, SSH3BP1 | |
Rabbit | |
Affinity Purified | |
RUO | |
10006 | |
Store at 4C. Do not freeze. |
Immunoprecipitation, Western Blot | |
1.0 mg/mL | |
Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative) | |
Q8IZP0 | |
ABI1 | |
The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human abl-interactor 1 using the numbering given in entry NP_005461.2 (GeneID 10006). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction